missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tungsten oxide hydrate

Product Code. 11440071
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440071 100 g 100g
11450071 25 g 25g
11460071 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440071 Supplier Thermo Scientific Alfa Aesar Supplier No. 044792.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12333-11-8 | (NH4)6W12O39·xH2O

Ammonium tungsten oxide hydrate is widely utilized as a raw material for the synthesis of tungsten catalysts, which is used in a variety of reactions like oxidation, hydroxylation, hydrogenation and polymerization. It is used as a reagent for chemical analysis like medical diagnosis. It is used as a precursor material for the preparation of high-purity WO3 which is used as thin-film substrate for certain semiconductor devices. It acts as a precursor to a variety of POM (polyoxometalates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12333-11-8
Molecular Formula (NH4)6W12O39·xH2O
MDL Number MFCD00042673
Synonym Ammonium metatungstate hydrate

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 2938.41 (Anhydrous)
Chemical Name or Material Ammonium tungsten oxide hydrate
compliance-icons
Product Identifier
  • Ammonium metatungstate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium tungsten oxide hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.