missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tungsten oxide, 99.99+% (metals basis)

Product Code. 11465370
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11465370

Brand: Thermo Scientific Alfa Aesar 044588.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11140-77-5 | H40N10O41W12 | 3042.429 g/mol

Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11140-77-5
Molecular Formula H40N10O41W12
Molecular Weight (g/mol) 3042.429
MDL Number MFCD00015956
InChI Key LMAJGSPZJYEENT-UHFFFAOYSA-D
Synonym ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine
PubChem CID 71306883
SMILES N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Soluble in warm water
Formula Weight 283.91
Percent Purity ≥99.99%
Assay (metals basis)
Chemical Name or Material Ammonium tungsten oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.