missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tungsten oxide, 99.99+% (metals basis)

Product Code. 11485370
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485370 250 g 250g
11465370 10 g 10g
11475370 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485370 Supplier Thermo Scientific Alfa Aesar Supplier No. 044588.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11140-77-5 | H40N10O41W12 | 3042.429 g/mol

Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11140-77-5
Molecular Formula H40N10O41W12
Molecular Weight (g/mol) 3042.429
MDL Number MFCD00015956
InChI Key LMAJGSPZJYEENT-UHFFFAOYSA-D
Synonym ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine
PubChem CID 71306883
SMILES N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O

Specifications

Physical Form Powder
Quantity 250 g
Solubility Information Soluble in warm water
Formula Weight 283.91
Percent Purity ≥99.99%
Assay (metals basis)
Chemical Name or Material Ammonium tungsten oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.