missing translation for 'onlineSavingsMsg'
Learn More

Ammonium trifluoromethanesulfonate, 99%

Product Code. 10646894 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Packaging:
Glass bottle
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646894 25 g Glass bottle 25g
10074923 5 g Glass bottle 5g
10608434 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10646894 Supplier Thermo Scientific Acros Supplier No. 388730250
  • ONLINE EXCLUSIVE PRICE
    £139.00
    £129.00 / 25g
    Save £10.00
    7% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38542-94-8 | CH4F3NO3S | 167.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38542-94-8
Molecular Formula CH4F3NO3S
Molecular Weight (g/mol) 167.11
MDL Number MFCD00075332
InChI Key BMWDUGHMODRTLU-UHFFFAOYSA-N
Synonym ammonium trifluoromethanesulfonate, ammonium triflate, acmc-20alsc, methanesulfonic acid,1,1,1-trifluoro-, ammonium salt 1:1
PubChem CID 15842047
IUPAC Name azanium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].[NH4+]

Specifications

Melting Point 225°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Formula Weight 167.11
Percent Purity 99%
Chemical Name or Material Ammonium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.