missing translation for 'onlineSavingsMsg'
Learn More

Ammonium trifluoroacetate, 98%

Product Code. 11446030
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446030 100 g 100g
11436030 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446030 Supplier Thermo Scientific Alfa Aesar Supplier No. H26577.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3336-58-1 | C2H5F3NO2 | 132.06 g/mol

Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of hexafluoro-2-aminopentan-4-one ligand and in tuning and calibrating new liquid chromatography/mass spectrometry systems. Applications of ammonium trifluoroacetate as an additive for elution of chiral acids and bases from derivatized polysaccharide stationary phases

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture. Keep under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
IUPAC Name azanium;2,2,2-trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F

Specifications

Melting Point 123°C to 126°C
Linear Formula CF3COONH4
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in Water.
Formula Weight 131.05
Percent Purity 98%
Chemical Name or Material Ammonium trifluoroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.