missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%, pure

Product Code. 10432180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10432180 2.5 kg Plastic bottle 2.5kg
10745642 500 g Plastic bottle 500g
10048329 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10432180 Supplier Thermo Scientific Acros Supplier No. 391660025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-18-8 | H8N2O3S2 | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-18-8
Molecular Formula H8N2O3S2
Molecular Weight (g/mol) 148.21
MDL Number MFCD00015965
Synonym ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

Specifications

Melting Point 150.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 95% min. (Iodimetry)
Linear Formula (NH4)2S2O3
Merck Index 15, 559
Solubility Information Solubility in water: 63.1% (20°C) - 71.0% (70°C). Other solubilities: insoluble in alcohol and ether
Formula Weight 148.21
Percent Purity 96%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium thiosulfate, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.