missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%, Thermo Scientific Chemicals

Catalog Number 11394599
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
7783-18-8
(NH4)2S2O3
MFCD00015965
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate
This item is not returnable. View return policy
7783-18-8
(NH4)2S2O3
MFCD00015965
ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

Solubility
Soluble in water, slightly soluble in acetone. Insoluble in ethanol and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
TRUSTED_SUSTAINABILITY
Melting Point ∼180°C (decomposition)
pH 6.0 to 6.5
Quantity 100 g
Merck Index 14,562
Solubility Information Soluble in water,slightly soluble in acetone. Insoluble in ethanol and diethyl ether.
Formula Weight 148.2
Percent Purity ≥96%
Sensitivity Hygroscopic
Density 1.64
Chemical Name or Material Ammonium thiosulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'