missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiosulfate, 96%

Product Code. 11439502
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439502 2500 g 2500g
11394599 100 g 100g
11429502 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439502 Supplier Thermo Scientific Alfa Aesar Supplier No. A10190.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-18-8 | (NH4)2S2O3

Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium thiosulfate finds application for leaching of gold and silver since it forms strong complexes. It is widely utilized as a photographic fixing agent, which acts quicklier than sodium thiosulfate fixers. It serves as a fertilizer. It acts as an additive to coal-waste mixtures to inhibit the formation of dangerous dioxins and furans. It is used as an analytical reagent in laboratories.

Solubility
Soluble in water, slightly soluble in acetone. Insoluble in ethanol and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids, magnesium and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-18-8
Molecular Formula (NH4)2S2O3
MDL Number MFCD00015965
Synonym ammonium thiosulphate, thiosulfuric acid h2s2o3 , diammonium salt, thiosulfuric acid h2s2o3 , ammonium salt 1:2, diammonium hypo, acmc-1bhnz, ammoniumthiosulfate, dsstox_cid_9703, dsstox_rid_78814, dsstox_gsid_29703, diammonium thiosulphate

Specifications

Melting Point ∼180°C (decomposition)
pH 6.0 to 6.5
Quantity 2500 g
Merck Index 14,562
Solubility Information Soluble in water,slightly soluble in acetone. Insoluble in ethanol and diethyl ether.
Formula Weight 148.2
Percent Purity ≥96%
Sensitivity Hygroscopic
Density 1.64
Chemical Name or Material Ammonium thiosulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.