missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate Solution 0.1M (0.1N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical™

Product Code. 10264290 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
6 AMP
Unit Size:
6 ampoules
Product Code. Quantity unitSize
10264290 6 AMP 6 ampoules
1 options
This item is not returnable. View return policy

Product Code. 10264290

Brand: Fisher Chemical J/1500C/90

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number 11428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

pH 4.5 to 6
Physical Form Liquid
Quantity 6 AMP
Formula Weight 76.12g/mol
Packaging Ampoule
compliance-icons
Product Identifier
  • Ammonium thiocyanate 0.1M solution (1M Concentrate ampoule)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.