missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate EU Pharmacopoeia 1M, Fisher Chemical™

Product Code. 12960594 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
Set of 200 mL + 800 mL
Packaging:
Bottle
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12960594 Set of 200 mL + 800 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12960594 Supplier Fisher Chemical Supplier No. RP2581000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Chemical Name or Material Ammonium molybdate solution R3 (Solution I + Solution II)
Quantity Set of 200 mL + 800 mL
Purity Grade Notes EUR Ph Ref: 1005704
Grade Eu. Ph.
Packaging Bottle
compliance-icons
Product Identifier
  • Ammonium Thiocyanate 1M Volumetric Solution (RP 258)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.