missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium thiocyanate, 99+%, extra pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
10kg
1kg
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 149.0°C |
| Color | White |
| pH | 4.0 to 6.0 (5% aq. soln.) |
| Physical Form | Crystals |
| Quantity | 10 kg |
| Assay Percent Range | 99% min. (on dried basis) (Argentometry) |
| Linear Formula | NH4SCN |
| Merck Index | 15,558 |
| Solubility Information | Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate |
| Formula Weight | 76.12 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur