missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 149.0°C |
| Color | White |
| pH | 4.0 to 6.0 (5% aq. soln.) |
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% min. (on dried basis) (Argentometry) |
| Linear Formula | NH4SCN |
| Merck Index | 15,558 |
| Solubility Information | Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol,acetone and ammonia,practically insoluble in chcl3 and ethyl acetate |
| Formula Weight | 76.12 |
| Show More |
Safety and Handling
Product Identifier
- Ammonium thiocyanate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Ammonium thiocyanate
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?