missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 0.1N Standardized Solution

Product Code. 11383677
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383677 4 L 4 L
11393677 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383677 Supplier Thermo Scientific Alfa Aesar Supplier No. 035580.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7732-18-5 | NH4SCN

Ammonium thiocyanate, 0.1N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula NH4SCN
MDL Number MFCD00011428

Specifications

Physical Form Liquid
Color Colorless
Solubility Miscible with water, ethanol, methanol, ammonia and acetone
Quantity 4 L
Formula Weight 76.12
Chemical Name or Material Ammonium thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.