missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Tetrathiomolybdate, TRC
Specifications
Specifications
| Chemical Name or Material | Ammonium Tetrathiomolybdate |
| Quantity | 1 g |
| Molecular Formula | MoS4 . 2(NH4) |
| Synonym | ATTM, Ammonium Molybdenum Sulfide, Ammonium Tetrasulfidomolybdate(2-), Ammonium Thiomolybdate, Bis(ammonium)tetrathiomolybdate(2-), Coprexa, Thiomolybdic Acid Diammonium Salt, Diammonium Tetrakis(sulfido)molybdate(2-), Diammonium Tetrakis(thioxo)molybdate, Diammonium Tetrasulfidomolybdate, Diammonium Tetrathiomolybdate, Diammonium Tetrathiomolybdate(2-), Diammonium Tetrathiooxomolybdate(2-), Diammonium Tetrathioxomolybdate(2-), Diammonium Thiomolybdate, Diammonium Thiomolybdate, NSC 286644, NSC 714598, TM, Tiomolibdate Diammonium |
| InChI Formula | InChI=1S/Mo.2H3N.4S/h;2*1H3;;;;/q;;;;;2*-1/p+2 |
| SMILES | [NH4+].[NH4+].[S-][Mo](=S)(=S)[S-] |
| IUPAC Name | diazanium;bis(sulfanylidene)-disulfidomolybdenum |
| Molecular Weight (g/mol) | 262.3 |
| Formula Weight | 261.8624 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?