missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrathiomolybdate, 99.95% (metals basis)

Product Code. 11375827
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375827 25 g 25g
11365827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375827 Supplier Thermo Scientific Alfa Aesar Supplier No. 043493.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15060-55-6 | H10MoN2S4 | 262.28 g/mol

Ammonium tetrathiomolybdate is used as an important reagent in synthetic chemistry and as a building block in bioinorganic chemistry. It is used in the treatment of copper toxicosis in animals and in Wilson's disease. It acts as a chelator in vivo to reduce copper levels. It interferes with angiogenesis and reduces tumor growth. Further, it is used in the synthesis of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands. It is involved in the ring opening of aziridines leading to sulfides and disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15060-55-6
Molecular Formula H10MoN2S4
Molecular Weight (g/mol) 262.28
MDL Number MFCD00136013
InChI Key ZKKLPDLKUGTPME-UHFFFAOYSA-N
Synonym ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
PubChem CID 15251598
IUPAC Name diazanium;bis(sulfanylidene)molybdenum;sulfanide
SMILES [NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S

Specifications

Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Slightly soluble in water.
Formula Weight 260.27
Sensitivity Hygroscopic
Chemical Name or Material Ammonium tetrathiomolybdate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.