missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 99%, pure

Product Code. 10369902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10369902 100 g Plastic bottle 100g
10393003 500 g Plastic bottle 500g
13438508 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10369902 Supplier Thermo Scientific Acros Supplier No. 310961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13826-83-0 | BF4H4N | 104.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
SMILES [NH4+].F[B-](F)(F)F

Specifications

Melting Point 230.0°C
Color Colorless
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula NH4BF4
Fieser 05,17; 06,132
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ammonium hydride
IUPAC Name azanium;tetrafluoroborate
Formula Weight 104.84
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium fluoborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.