missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium tetrafluoroborate, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Ammonium fluoroborate is used as an analytical reagent. It is also applied in the textile printing and dyeing industry and used as a catalyst for resin finishing. It is mainly used for the high temperature fluxing action required by the metals industry; catalyst; in flame retardants and acts as a solid lubricant in cutting-oil emulsions for aluminum rolling and forming.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220°C (sublimation) |
Color | White |
Physical Form | Crystalline |
Quantity | 5 g |
UN Number | UN3260 |
Solubility Information | Soluble in water at 16°C 25g/L. |
Formula Weight | 104.84 |
Percent Purity | 99.999% |
Odor | Ammonia-like |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'