missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrafluoroborate, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11348727
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ammonium tetrafluoroborate is used in the metal industry as a high-temperature flux. It is also used as a flame retardant and lubricant in aluminum cutting-oils. Further, it acts as an analytical reagent. In addition, it is used as a catalyst for resin finishing and also finds use in textile printing and dyeing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13826-83-0
Molecular Formula BF4H4N
Molecular Weight (g/mol) 104.84
MDL Number MFCD06245785
InChI Key PDTKOBRZPAIMRD-UHFFFAOYSA-O
Synonym ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
PubChem CID 9964072
IUPAC Name azanium;tetrafluoroborate
SMILES [NH4+].F[B-](F)(F)F

Specifications

Melting Point 220°C (sublimation)
Color White
Quantity 1 kg
UN Number UN3260
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 104.84
Percent Purity 99.5%
Odor Ammonia-like
Sensitivity Hygroscopic
Density 1.871 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'