missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min

Product Code. 11341688
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341688 5 g 5g
11351688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341688 Supplier Thermo Scientific Alfa Aesar Supplier No. 011046.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 13820-41-2 | Cl4H8N2Pt | 372.96 g/mol

Ammonium tetrachloroplatinate(II) is used in spectral analysis standard. It is also used in the preparation of platinum sponge and platinum catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-41-2
Molecular Formula Cl4H8N2Pt
Molecular Weight (g/mol) 372.96
MDL Number MFCD00010885
InChI Key QJIMNDWDOXTTBR-UHFFFAOYSA-L
Synonym ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
IUPAC Name platinum(2+) diammonium tetrachloride
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 145°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 5 g
Linear Formula (NH4)2PtCl4
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 372.98
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 51% min.
Odor Odorless
Density 2.936 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.