missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Ammonium tetrachloropalladate(II) is one of the preferred complexes, which are used for the preparation of palladium metallic particles. In addition, it forms Pd-creatinine complexes, which find use in the field of catalysis, and semiconductors. Colloidal palladium nanocatalysts and metal polymer catalyst systems can be prepared from ammonium tetrachloropalladate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Brown to Green |
Physical Form | Powder |
Quantity | 1 g |
Linear Formula | (NH4)2PdCl4 |
Solubility Information | Soluble in water |
Formula Weight | 284.31 |
Percent Purity | 99.998% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Pd 37% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'