missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloropalladate(II), Pd 36% min

Product Code. 11331968
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331968 1 g 1g
11341968 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331968 Supplier Thermo Scientific Alfa Aesar Supplier No. 011882.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-40-1 | Cl4H8N2Pd | 284.30 g/mol

It is used as catalyst, and as a standard for spectral analysis. It reacts with creatinine to form various Pd-creatinine complexes which are used in semiconductors, catalysis, mixed oxidation state studies, and electron conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-40-1
Molecular Formula Cl4H8N2Pd
Molecular Weight (g/mol) 284.30
MDL Number MFCD00010884
InChI Key IRKVTUVFHGUMMN-UHFFFAOYSA-L
Synonym ammonium tetrachloropalladate ii, ammonium palladium chloride, diammonium tetrachloropalladate 2-, ammonium chloropalladite, bisammonium tetrachloropalladate 2-, diammonium tetrachloro palladite, ammonium tetrachloropalladate, ccris 7828, palladate 2-, tetrachloro-, diammonium, sp-4-1, ammoniumpalladiumchloride
PubChem CID 61682
ChEBI CHEBI:60149
IUPAC Name diazanium;tetrachloropalladium(2-)
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]

Specifications

Melting Point (decomposition)
Color Brown to Green
Physical Form Powder
Quantity 1 g
Linear Formula (NH4)2PdCl4
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 284.31
Concentration or Composition (by Analyte or Components) Pd 36% min.
Sensitivity Hygroscopic
Density 2.17 g/mL
Chemical Name or Material Ammonium tetrachloropalladate(II)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium tetrachloropalladate(II)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium chloropalladite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.