missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis)

Product Code. 11368388
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368388 1 g 1g
11358388 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368388 Supplier Thermo Scientific Alfa Aesar Supplier No. 040434.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13874-04-9 | AuCl4H4N | 356.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13874-04-9
Molecular Formula AuCl4H4N
Molecular Weight (g/mol) 356.82
MDL Number MFCD00011503
InChI Key APXMUHKEDBIQKQ-UHFFFAOYSA-J
Synonym ammonium tetrachloroaurate iii hydrate, ammonium tetrachloroaurate iii hydrate, premion, ammonium tetrachloroaurate ii, azanium;tetrachlorogold 1-;hydrate, ammonium tetrachloroaurate iii n-hydrate, ammonium tetrachloroaurate 1--water 1/1/1, ammoniumtetrachloroaurate iii hydrate-au puratrem, ammonium tetrachloroaurate iii hydrate, premion r metals basis
PubChem CID 16211474
SMILES N.Cl[Au](Cl)(Cl)Cl

Specifications

Color Yellow to Orange
Physical Form Crystalline Powder
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in water and alcohol
IUPAC Name azanium;tetrachlorogold(1-);hydrate
Formula Weight 356.82 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloroaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.