missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Ammonium Test

Brand:  Macherey-Nagel™ 90722


Product Code. 11346524

  • ONLINE EXCLUSIVE PRICE
    £50.30
    £46.81 / Pack of 200
    Save £3.49
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

White to Brown/Yellow
Determination of ammonium and ammonia
pH Paper
Strip
200 Pack
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ammonium test paper

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : potassium tetraiodomercurate(II)

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.