missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Tartrate Dibasic, purum p.a., ≥99.0% (T), Honeywell™

Product Code. 15620940 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15620940 250 g Poly Bottle 250g
15610940 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15620940 Supplier Honeywell Research Chemicals Supplier No. 09984250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a., crystallized,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
IUPAC Name diazanium;(2R,3R)-2,3-dihydroxybutanedioate
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

Color White
Physical Form Crystallized
Quantity 250 g
Linear Formula (NH4)2C4H4O6
Beilstein 6120352
Formula Weight 184.15g/mol
Percent Purity ≥99.0% (T)
Grade purum p.a.
Packaging Poly Bottle
Residue after Ignition ≤0.05% (as SO4)
Appearance Crystals
Density 1.601g/cu.cm
Chemical Name or Material Ammonium Tartrate Dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.