missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfide Solution, 40 to 48 wt.% in H2O, Honeywell™ Fluka™

Product Code. 15635730 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15635730 500 mL Glass Bottle 500mL
15615730 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15635730 Supplier Honeywell-Fluka Supplier No. 515809500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Boiling Point 40°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Linear Formula (NH4)2S
UN Number UN2683
Flash Point 20°C (68°F)
Formula Weight 68.14g/mol
Concentration or Composition (by Analyte or Components) 40 to 48 wt. % in H2O
Packaging Glass Bottle
Density 1g/mL at 25°C
Chemical Name or Material Ammonium sulfide solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.