missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 21.0%, MP Biomedicals™

Product Code. 11496251 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496251 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496251 Supplier MP Biomedicals Supplier No. 0219386080
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 12135-76-1
Color Yellow
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 0.21
Molecular Formula H8N2S
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
SMILES N.N.S
IUPAC Name diamine sulfane
Molecular Weight (g/mol) 68.14
Formula Weight 68.1
Density 0.997g/mL
Chemical Name or Material Ammonium sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.