missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 20% solution in water

Product Code. 10515524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10515524 1 L Glass bottle 1 L
10490694 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515524 Supplier Thermo Scientific Acros Supplier No. 426080010
  • ONLINE EXCLUSIVE PRICE
    £277.00
    £241.65 / 1 L
    Save £35.35
    12% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12135-76-1 | H8N2S | 68.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Quantity 1 L
Linear Formula (NH4)2S
Flash Point 32°C
Merck Index 15, 553
Solubility Information Solubility in water: miscible
Formula Weight 68.14
Concentration or Composition (by Analyte or Components) 20.0 to 24.0% (Iodimetry)
Packaging Glass bottle
Appearance Clear yellow solution
Specific Gravity 1
Density 1.0000g/mL
Chemical Name or Material Ammonium sulfide, 20% solution in water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ammonium sulfide, 20% solution in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.