missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 20-24% aq. soln.

Product Code. 11454990
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454990 500 mL 500mL
11347598 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454990 Supplier Thermo Scientific Alfa Aesar Supplier No. 033286.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 12135-76-1 | H8N2S | 68.14 g/mol

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth. It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S

Specifications

Color Colorless to Orange
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 20 to 24% (aqueous)
Flash Point 32°C (90°F)
Solubility Information Miscible with water,alcohol and liquid ammonia.
Formula Weight 68.14
Odor Like rotten eggs
Appearance Colorless liquid
Vapor Pressure 546 hPa (410mm Hg) at 20°C
Density 0.997 g/cm3 at 20°C
Chemical Name or Material Ammonium sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.