missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultra Pure, 25kg, MP Biomedicals

Product Code. 11411642
Click to view available options
Quantity:
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Fe: <<0.5 ppm
Mg: <<0.13 ppm
Pb: <<0.1 ppm
Mn: <<0.06 ppm
Ca: <<0.1 ppm
Zn: <<0.5 ppm
Cu: <<0.21 ppm
O.D. (260nm) of a saturated solution: <<0.1

TRUSTED_SUSTAINABILITY

Specifications

CAS 7783-20-2
Quantity 25 kg
Molecular Formula H8N2O4S
MDL Number MFCD00003391
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILES N.N.OS(O)(=O)=O
IUPAC Name diazanium;sulfate
Molecular Weight (g/mol) 132.13
PubChem CID 6097028
ChEBI CHEBI:62946
Grade Ultrapure
Chemical Name or Material Ammonium Sulfate, Ultra Pure
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'