missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
50 kg
Unit Size:
1kg
50kg
5kg
Pack of 4
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 5 kg |
| Linear Formula | (NH4)2SO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 132.14g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Residue after Ignition | ≤0.005% (as SO4) |
| Density | 1.77g/mL at 25°C |
| Chemical Name or Material | Ammonium Sulfate, Reag. Ph. Eur. reag. ISO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction