missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, ACS, 99.0% min

Product Code. 11327297
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
Unit Size:
100g
10kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327297 10 kg 10kg
11337297 100 g 100g
11347297 500 g 500g
11357297 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11327297 Supplier Thermo Scientific Alfa Aesar Supplier No. 011566.A9
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

In galvanizing steel, manufacturing of ammonia alum and sulfuric acid | CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. Ammonium sulfate is used in agriculture as fertilizer. It is a slow releasing nitrogenous fertilizer and a rich source of sulphur nutrient. It is employed as a dough conditioner in bread products. It finds use as raw materials for various chemical products such as dyes and pharmacetical products. It is a component of flame-proofing agents and fire extinguisher powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point ∽280°C (decomposition)
Color White
Physical Form Granules
Quantity 10 kg
Assay Percent Range 99% min.
Solubility Information Highly soluble in water. It is insoluble in acetone,ether and ethanol. It is also insoluble in liquid ammonia.
IUPAC Name sulfuric acid diamine
Formula Weight 132.14
Grade ACS
Appearance White granular
Density 1.769
Chemical Name or Material Ammonium sulfate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.