missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, 99+%, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2.5kg
5kg
Specifications
Specifications
| Melting Point | 280°C |
| Color | Colorless |
| pH | 5 to 6 |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Formula Weight | 132.13g/mol |
| Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction