missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10171940 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2.5kg
5kg
This item is not returnable. View return policy

Product Code. 10171940

Brand: Fisher Chemical A/6440/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Melting Point 280°C
Color Colorless
pH 5 to 6
Physical Form Solid
Quantity 5 kg
Formula Weight 132.13g/mol
Packaging Plastic keg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.