missing translation for 'onlineSavingsMsg'
Learn More

Melford Ammonium Sulfate

Product Code. 30018004
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018004 5 kg 5kg
30018003 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018004 Supplier Melford Supplier No. A205105000.0
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Useful for precipitation or fractionation of proteins or for purification of antibodies.

Useful for precipitation or fractionation of proteins or for purification of antibodies. Used in crystallographic analysis of nucleic acids and proteins.

Specifications

Chemical Name or Material Ammonium Sulfate
CAS 7783-20-2
Quantity 5 kg
Molecular Formula H8N2O4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.