missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfamate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.
Solubility
Soluble in water(2800g/L ). Insoluble in methanol, ether, n-octanol.
Notes
Hygroscopic. Store at room temperature. Stored in airtight containers which are kept closed and away from air and moisture.
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.
Solubility
Soluble in water(2800g/L ). Insoluble in methanol, ether, n-octanol.
Notes
Hygroscopic. Store at room temperature. Stored in airtight containers which are kept closed and away from air and moisture.
Specifications
Boiling Point | 160°C (decomposition) |
Melting Point | 131°C to 135°C |
pH | 5.2 |
Quantity | 250 g |
Linear Formula | NH4SO3NH2 |
Merck Index | 14,554 |
Solubility Information | Soluble in water(2800g/L ). Insoluble in methanol,ether,n-octanol. |
Formula Weight | 114.12 |
Percent Purity | ≥98% |
Odor | Weak |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'