missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sodium phosphate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10461261
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-13-3
Molecular Formula H5NNaO44H2O
Molecular Weight (g/mol) 209.07
MDL Number MFCD00066970
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
PubChem CID 56924540
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

Specifications

Melting Point 80°C
Color Colorless to White
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Assay Percent Range 99%
Linear Formula Na(NH4)HPO4·4H2O
Merck Index 12,587
Formula Weight 209.07
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.54
Density 1.5400g/mL
Chemical Name or Material Ammonium sodium phosphate tetrahydrate, For analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'