missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sodium Phosphate Dibasic Tetrahydrate, Honeywell Fluka™

Product Code. 15670730 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
Unit Size:
1kg
500g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15670730 500 g 500g
15660730 1 kg 1kg
15690730 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15670730 Supplier Honeywell-Fluka Supplier No. 04266500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-13-3
Molecular Formula H13NNaO8P
Molecular Weight (g/mol) 209.067
MDL Number MFCD00066970
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
PubChem CID 56924540
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

Specifications

Quantity 500 g
Linear Formula NaNH4HPO4 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 209.07g/mol
Percent Purity ≥99%
Grade puriss.
Chemical Name or Material Ammonium Sodium Phosphate Dibasic Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.