missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sodium Hydrogen Orthophosphate Tetrahydrate, Extra Pure, SLR, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
Specifications
Specifications
| Melting Point | 79°C |
| CAS | 13011-54-6 |
| Color | Colorless |
| pH | 8 |
| Physical Form | Solid |
| Quantity | 500 g |
| Molecular Formula | H5NNaO4P |
| MDL Number | 66970 |
| InChI Key | CUXQLKLUPGTTKL-UHFFFAOYSA-M |
| Molecular Weight (g/mol) | 137.01 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction