missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Pyrrolidine Dithiocarbamate, for AAS, Fisher Chemical

Product Code. 10746091 Shop All Fisher Chemical Products
Change view
Click to view available options
25g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10746091 25g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10746091 Supplier Fisher Chemical Supplier No. A/6220/46
Quantity Price
16 £159.48 / 25g
1 £187.63 / 25g
4 £168.87 / 25g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 150°C
CAS 5108-96-3
Color White
pH 6 to 7
Physical Form Powder/Solid
Quantity 25 g
Molecular Formula C5H12N2S2
MDL Number 12720
Formula Weight 164.28g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • 1-Pyrrolidinecarbodithioic acid, ammonium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.