missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate (Monobasic), Fisher BioReagents

Product Code. 10744914
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10744914

Brand: Fisher BioReagents BP2427500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Ammonium Phosphate, Monobasic is used in several biochemical research applications.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 190°C
CAS 7722-76-1
Color White
pH 3.8 to 4.4
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98.0 %
Molecular Formula H6NO4P
Linear Formula (NH4)H2PO4
MDL Number MFCD00003396
Merck Index 15, 541
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES N.OP(O)(O)=O
IUPAC Name phosphoric acid amine
Molecular Weight (g/mol) 115.03
PubChem CID 24402
ChEBI CHEBI:62982
Formula Weight 115.03
Percent Purity ≥98.0%
Identification Pass Test
Packaging Glass Bottle
Decomposition Information >190°C
Chemical Name or Material Ammonium Phosphate, Monobasic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.