missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic, ≥99.99% Trace Metals Basis, Honeywell™

Product Code. 15655620 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 15655620

Brand: Honeywell Research Chemicals 467782250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Boiling Point 197°C
Melting Point 190°C
Color Colorless
pH 3.8 to 4.4 (at 50g/L 25°C)
Quantity 250 g
Linear Formula NH4H2PO4
UN Number NONH for all modes of transport
Formula Weight 115.03g/mol
Percent Purity ≥99.99% (trace metals basis)
Odor Odorless
Packaging Poly Bottle
Appearance Solid
Chemical Name or Material Ammonium phosphate monobasic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.