missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, TRC

Product Code. 30341904
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341904 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341904 Supplier Toronto Research Chemicals Supplier No. TRCA634065100G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Quantity 100 g
Molecular Formula H8N2O8S2
Synonym Peroxydisulfuric Acid ([(HO)S(O)2]2O2) Ammonium Salt (1:2), Peroxydisulfuric Acid ([(HO)S(O)2]2O2) Diammonium Salt, Ad 485, Ammonium Peroxidodisulfate, Ammonium Peroxydisulfate, Ammonium Peroxydisulfate ((NH4)2S2O8), Ammonium Peroxysulfate, Anala R, Bis(ammonium) Peroxodisulfate, Diammonium Peroxydisulfate, Diammonium Peroxydisulphate, Diammonium Persulfate, Enplate AD 485, Panreac PA
InChI Formula InChI=1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
IUPAC Name diazanium;sulfonatooxy sulfate
Molecular Weight (g/mol) 228.2
Formula Weight 227.9722
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.