missing translation for 'onlineSavingsMsg'
Learn More

Melford Ammonium Persulfate [APS]

Product Code. 30018001
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018001 100 g 100g
30018002 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018001 Supplier Melford Supplier No. A20500100.0
  • ONLINE EXCLUSIVE PRICE
    £73.10
    £44.00 / 100g
    Save £29.10
    39% Off
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a catalyst for acrylamide gel polymerization.

Used as a catalyst for acrylamide gel polymerization.
  • Heavy Metals: <0.005%

Specifications

Chemical Name or Material Ammonium Persulfate [APS]
CAS 7727-54-0
Quantity 100 g
Molecular Formula H8N2O8S2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.