missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, ACS, ≥98%, Ultrapure

Product Code. 15835378 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
10g
1kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835378 1 kg Plastic bottle 1kg
15885228 10 g Plastic bottle 10g
15895228 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15835378 Supplier Thermo Scientific Supplier No. J76322.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High speed initiator used to catalyze acrylamide polymerization. | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

A high speed initiator used to catalyze acrylamide polymerization.

Working Stock Solution (10%):

  • Ammonium Persulfate - 1 gm
  • dH2O to 10 ml
  • This solution should be stored at 4°C and prepared fresh weekly.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline Powder
Quantity 1 kg
UN Number UN1444
Formula Weight 228.19
Percent Purity ≥98%
Grade ACS
Packaging Plastic bottle
Specific Gravity 1.982
Density 1.982
Chemical Name or Material Ammonium Persulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.