missing translation for 'onlineSavingsMsg'
Learn More

Ammonium persulfate, 99+%, for molecular biology, DNAse, RNAse and Protease free

Product Code. 10024273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10024273 500 g Plastic bottle 500g
10769081 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10024273 Supplier Thermo Scientific Acros Supplier No. 327085000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Melting Point 120.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula (NH4)2S2O8
Fieser 01,952; 02,348; 03,238; 05,15; 06,20; 12,33
Merck Index 15, 539
Solubility Information Solubility in water: 582g/L (20°C),may decompose
Formula Weight 228.19
Percent Purity ≥99%
Grade Molecular Biology
Packaging Plastic bottle
Specific Gravity 1.98
Density 1.9800g/mL
Chemical Name or Material Ammonium persulfate, DNAse, RNAse and Protease Free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium persulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.