missing translation for 'onlineSavingsMsg'
Learn More

Ammonium perrhenate(VII), Puratronic™, 99.999% (metals basis)

Product Code. 11316997
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316997 2 g 2g
11326997 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316997 Supplier Thermo Scientific Alfa Aesar Supplier No. 010846.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13598-65-7 | H4NO4Re | 268.24 g/mol

Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-65-7
Molecular Formula H4NO4Re
Molecular Weight (g/mol) 268.24
MDL Number MFCD00012377
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
Synonym azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
PubChem CID 3084163
SMILES N.O[Re](=O)(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline
Quantity 2 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water.
IUPAC Name azanium;oxido(trioxo)rhenium
Formula Weight 268.24
Appearance White crystalline
Density 3.97 g/cm3
Chemical Name or Material Ammonium perrhenate(VII)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.