missing translation for 'onlineSavingsMsg'
Learn More

Ammonium perrhenate(VII), 99+% (metals basis), Thermo Scientific Chemicals

Catalog Number 11351219
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
13598-65-7
H4NO4Re
268.24
MFCD00012377
XPSXHXWHJAIQJR-UHFFFAOYSA-M
azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
3084163
N.O[Re](=O)(=O)=O
This item is not returnable. View return policy
13598-65-7
H4NO4Re
268.24
MFCD00012377
XPSXHXWHJAIQJR-UHFFFAOYSA-M
azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
3084163
N.O[Re](=O)(=O)=O

Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 2 g
Linear Formula NH4ReO4
UN Number UN1479
Solubility Information Soluble in water.
IUPAC Name ammonium tetraoxorheniumuide
Formula Weight 268.24
Percent Purity ≥99%
Density 3.97 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium perrhenate(VII)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'