missing translation for 'onlineSavingsMsg'
Learn More

Ammonium perrhenate(VII), 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11361219
Click to view available options
Quantity:
2 g
10 g
50 g

Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13598-65-7
Molecular Formula H4NO4Re
Molecular Weight (g/mol) 268.24
MDL Number MFCD00012377
InChI Key XPSXHXWHJAIQJR-UHFFFAOYSA-M
Synonym azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis
PubChem CID 3084163
SMILES N.O[Re](=O)(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula NH4ReO4
UN Number UN1479
Solubility Information Soluble in water.
IUPAC Name azanium;oxido(trioxo)rhenium
Formula Weight 268.24
Percent Purity ≥99%
Density 3.97 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium perrhenate(VII)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'