missing translation for 'onlineSavingsMsg'
Learn More

Ammonium pentaborate tetrahydrate, 98%

Product Code. 15442667
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442667 2 kg 2kg
15432667 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442667 Supplier Thermo Scientific Alfa Aesar Supplier No. 046323.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12046-04-7 | B5H44N9O19-6 | 528.446 g/mol

Ammonium pentaborate is used in the preparation of both wet and dry electrolytic capacitors, flame proofing, gunning and patching compounds for extending the life of basic refractories. Used in the preparation of special welding, soldering and brazing fluxes for stainless steel or various non-ferrous metals, where alkali borates cannot be used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12046-04-7
Molecular Formula B5H44N9O19-6
Molecular Weight (g/mol) 528.446
MDL Number MFCD13176621
InChI Key OJAJAQRDJBKPPU-UHFFFAOYSA-W
Synonym nonaammonium tetrahydrate pentaborate
PubChem CID 57347064
IUPAC Name nonaazanium;pentaborate;tetrahydrate
SMILES B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].B([O-])([O-])[O-].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].[NH4+].O.O.O.O

Specifications

Melting Point 110°C
Color White
Physical Form Powder
Quantity 2 kg
Linear Formula (NH4)2B10O16·4H2O
Solubility Information Soluble only in water.
Formula Weight 272.15 (200.09 Anhydrous)
Percent Purity ≥99%
Density 1.58 g/mL
Chemical Name or Material Ammonium pentaborate tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.