Learn More
Ammonium pentaborate tetrahydrate, 98%
Description
Ammonium pentaborate is used in the preparation of both wet and dry electrolytic capacitors, flame proofing, gunning and patching compounds for extending the life of basic refractories. Used in the preparation of special welding, soldering and brazing fluxes for stainless steel or various non-ferrous metals, where alkali borates cannot be used.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 110°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 500 g |
| Linear Formula | (NH4)2B10O16·4H2O |
| Solubility Information | Soluble only in water. |
| Formula Weight | 272.15 (200.09 Anhydrous) |
| Percent Purity | ≥99% |
| Density | 1.58 g/mL |
| Chemical Name or Material | Ammonium pentaborate tetrahydrate |
Safety and Handling
- Ammonium pentaborate tetrahydrate
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: Ammonium pentaboratetetrahydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.